Learn More
3M™ Versaflo™ TR-600/800 PAPR Accessory, Cleaning and Storage Kit TR-653 / 37373 (AAD)
Protects from internal contamination during cleaning and storage.
Brand: 3M™ 7100035841
49.25 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.06000kg
Includes: 1 inlet plug and 2 seals, 1 outlet plug and 2 seals and 1 battery connector cover
Description
- Plug and seal for inlet and outlet openings of TR-600 series motor/blower
- Cover for electrical connections of TR-630 or TR-632 battery
- Motor/blower plugs allow motor/blower to be submerged, rinsed under running water, or machine washed
- Plug seals to be replaced after 30 uses or whenever found to be damaged or worn
- 3M Catalog# TR-653
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Pesticide Application, Heavy Industrial, Animal processing, Laboratories, Nuclear energy, Manufacturing, Cosmetics, Pharmaceutical research and production, Recycling, General Manufacturing, Dry Foods
Certifications
NIOSH Approved
Specifications
Reusable | |
TR-600/TR-800 PAPR System | |
TR-653 |
Cleaning and Storage Plug | |
1 inlet plug and 2 seals, 1 outlet plug and 2 seals, and 1 battery connector cover | |
1 Ea. |