Learn More
3M™ Versaflo™ TR-600/800 Series PAPR Accessory, Battery Charger Cradle TR-640
Provides easy-to-use interface between all TR-600 Series batteries and all TR-900 Series power supplies.
Brand: 3M™ 7000144503
129.44 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement part for TR-641 and TR-644 battery charger kits
- Holds TR-630 or TR-632 batteries
- Can be used on TR-944 4 station power supply
- 3M Catalog# TR-640
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
Specifications
Single-Unit Charging Station | |
TR-640 | |
1/Cs. |
TR-600/800 Series PAPR Systems | |
One |