Learn More
3M™ Versaflo™ TR-600 Series PAPR Accessory, Prefilter
For use with Versaflo™ TR-600 Series PAPR
Brand: 3M™ 7100021255
15.75 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00360kg
Description
- TR-6600 prefilter extends life of particulate (HE) element of TR-600 series filters and cartridges by capturing large particles that may otherwise clog HE filter
- Frequent prefilter changeout optimizes PAPR system performance
- Use of the TR-6600 may reduces PAPR operating costs
- Easy to use due to simple maintenance free design
- Flexible application as TR-6600 may be used with TR-662 spark arrestor
- Black
- 3M Catalog# TR-6600
Applications:
Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
Specifications
Black | |
Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing | |
TR-6600 |
TR-6600 Prefilter | |
Versaflo™ TR-600 series filters and cartridges | |
50/Cs. |