Learn More
3M™ Versaflo™ TR-600 Series PAPR Accessory, Spark Arrestor
For use with Versaflo™ TR-600 Series PAPR.
Brand: 3M™ 7100035845
14.40 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- TR-662 spark arrestor is used with all TR-600 series filters and cartridges
- Protects TR-6000 series filters and cartridges from potential damage due to sparks
- Acts as pre-filter by capturing larger particles that would load main filter
- Flexible application as TR-662 may be used with TR-6600 prefilter
- Reduces operating costs by extending life of TR-6000 series filters and cartridges
- Color: Silver
- 3M Catalog# TR-662
Applications:
Healthcare
Certifications
NIOSH Approved
Specifications
Silver | |
Spark Arrestor | |
Versaflo™ TR-600 Series PAPR System | |
2/Cs. |
Reusable | |
Healthcare | |
TR-662 |