Learn More
3M™ Versaflo™TR-600 Series Filter Covers
Flter cover for use with TR-6320N, TR-6350N and TR-6360N cartridges
Brand: 3M™ 7100021336
11.60 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.20000kg
Description
- Helps protect cartridge from physical damage
- Holds TR-6600 Prefilter or TR-662 Spark Arrestor in place
- Enables PAPR to be worn through decontamination showers or rain
- Easy to use
- 3M Catalog# TR-6300FC
Applications:
Recommended Industry: Healthcare, Pharmaceutical
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
Filter Cover TR-6300FC / 37380 (AAD) | |
TR-6300 Series Cartridges | |
TR-6300FC | |
1/Cs. |