Learn More
STEMCELL Technologies 3T3-J2 Irradiated Feeder Cells
Product Code. 17141046
Encompass
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.50mL
Product Code. 17141046
Brand: STEMCELL Technologies 1000353
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Irradiated 3T3-J2 mouse fibroblast, frozen
  • 3T3-J2 Irradiated Feeder Cells are optimized to support epithelial cell expansion in Conditional Reprogramming (CR) Medium (Catalog #100-0352; Liu X et al. 2012; Liu X et al. 2017; Suprynowicz FA et al.) The cells are mitotically inactivated by radiation, but maintain metabolic activity
  • 3T3-J2 Irradiated Feeder Cells are cryopreserved and can be used directly for seeding epithelial cells without plating in advance
  • Each vial contains 2 - 3 x 10^6 cells and can be used to seed up to three T-25 cm2 flasks or one T-75 cm2 flask
  • 3T3-J2 Irradiated Feeder Cells are intended for Research Use Only
Specifications
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 0.5 mL