Learn More
Viking Optical™ 3X Hand Magnifier
Brand:  Viking Optical™ 5003
Encompass_Preferred
Product Code. 12866335
Description
Description
Bi-convex glass lens in metal rim with black plastic handle.
Specifications
Specifications
Bi-convex glass lens in metal rim with black plastic handle
76 mm
3X
Hand Magnifier
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Viking Optical™ 3X Hand Magnifier