missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,3-Thiadiazol-4-yl)benzamide, 95%, Thermo Scientific™

Product Code. 10324202
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10324202

Brand: Thermo Scientific Maybridge SEW01125DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-53-5
Molecular Formula C9H7N3OS
Molecular Weight (g/mol) 205.235
MDL Number MFCD00173809
InChI Key PAVNUEROKSNSDS-UHFFFAOYSA-N
Synonym 4-1,2,3-thiadiazol-4-yl benzamide, maybridge1_007554, 4-thiadiazol-4-yl benzamide, benzamide,4-1,2,3-thiadiazol-4-yl
PubChem CID 2739359
IUPAC Name 4-(thiadiazol-4-yl)benzamide
SMILES C1=CC(=CC=C1C2=CSN=N2)C(=O)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.24
Percent Purity 95%
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.