missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10600105
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 160068-88-2
Molecular Formula C10H11BO3
Molecular Weight (g/mol) 190.005
MDL Number MFCD04039026
InChI Key AMQBUMNOSGJNQL-UHFFFAOYSA-N
Synonym 4-1,3,2-dioxaborinan-2-yl benzaldehyde, 4-formylbenzeneboronic acid, propane-1,3-diol cyclic ester, 4-formylbenzeneboronic acid,propane-1,3-diol cyclic ester, 4-1,3,2-dioxaboran-2-yl benzaldehyde, 2-4-formylphenyl-1,3,2-dioxaborinane
PubChem CID 2794711
IUPAC Name 4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
SMILES B1(OCCCO1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 190
Percent Purity 97%
Chemical Name or Material 4-(1, 3, 2-Dioxaborinan-2-yl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.