missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10699505
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10699505

Brand: Thermo Scientific Maybridge MO01234DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 160068-88-2
Molecular Formula C10H11BO3
Molecular Weight (g/mol) 190.005
MDL Number MFCD04039026
InChI Key AMQBUMNOSGJNQL-UHFFFAOYSA-N
Synonym 4-1,3,2-dioxaborinan-2-yl benzaldehyde, 4-formylbenzeneboronic acid, propane-1,3-diol cyclic ester, 4-formylbenzeneboronic acid,propane-1,3-diol cyclic ester, 4-1,3,2-dioxaboran-2-yl benzaldehyde, 2-4-formylphenyl-1,3,2-dioxaborinane
PubChem CID 2794711
IUPAC Name 4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
SMILES B1(OCCCO1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 190
Percent Purity 97%
Chemical Name or Material 4-(1, 3, 2-Dioxaborinan-2-yl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.