missing translation for 'onlineSavingsMsg'
Learn More
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech., Thermo Scientific™ Product Code.: 15430656

4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech., Thermo Scientific™

Product Code. 15430656
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15430656

Brand: Thermo Scientific Alfa Aesar H53499.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190059-33-7
Molecular Formula C13H20O2
Molecular Weight (g/mol) 208.30
MDL Number MFCD07784337
InChI Key ODMUHAHUBCUABS-UHFFFAOYNA-N
Synonym 4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-3-buten-2-one, 4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 heptan-2-yl but-3-en-2-one, 3-buten-2-one,4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-, 3e, 4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-3-buten-2-one, mixture of cis and trans
PubChem CID 11041982
SMILES CC(=O)C=CC1C2(C)OC2CCC1(C)C

Specifications

Density 1.039
Refractive Index 1.491
Quantity 25 g
IUPAC Name (E)-4-(4,4,6-trimethyl-7-oxabicyclo[4.1.0]heptan-5-yl)but-3-en-2-one
Formula Weight 208.3
Grade Technical
Chemical Name or Material 4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM23A041 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.