missing translation for 'onlineSavingsMsg'
Learn More
4-(1-Cyclohexen-1-yl)morpholine, 97%, Thermo Scientific Chemicals Product Code.: 11449487

4-(1-Cyclohexen-1-yl)morpholine, 97%, Thermo Scientific Chemicals

Product Code. 11449487
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11449487

Brand: Thermo Scientific Alfa Aesar B22004.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-80-4
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00006163
InChI Key IIQFBBQJYPGOHJ-UHFFFAOYSA-N
Synonym 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene
PubChem CID 69589
IUPAC Name 4-(cyclohexen-1-yl)morpholine
SMILES C1CCC(=CC1)N2CCOCC2

Specifications

Density 1.02
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.5146
Quantity 25 g
Beilstein 118696
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 4-(1-Cyclohexen-1-yl)morpholine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10233499 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberB22004.22,B22004.14
Lot Number10135514 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22004.22,B22004.14
Lot Number10233499 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22004.22,B22004.14
Lot Number10024969 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22004.22,B22004.14
Lot Number10185255 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22004.22,B22004.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1-Cyclohexen-1-yl)morpholine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.