missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15479145
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938043-31-3
Molecular Formula C18H26BNO3
Molecular Weight (g/mol) 315.22
MDL Number MFCD09027083
InChI Key YOXMAOLZXRIELZ-UHFFFAOYSA-N
Synonym 4-piperidine-1-carbonyl phenylboronic acid pinacol ester, piperidin-1-yl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-1-piperidinylcarbonyl benzeneboronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperidine, 4-piperidine carboxamidophenylboronic acid, pinacol ester, amtb002, 4-piperidine-1-carbonyl phenylboronic acid, pinacol ester, 4-piperidine-1-carbonyl phenyl boronic acid pinacol ester
PubChem CID 16218322
IUPAC Name piperidin-1-yl-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)N3CCCCC3

Specifications

Quantity 1 g
Formula Weight 315.21
Chemical Name or Material 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.