missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11416886
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2456-81-7
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD00006415
InChI Key RGUKYNXWOWSRET-UHFFFAOYSA-N
Synonym 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
PubChem CID 75567
IUPAC Name 4-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=NC=C2

Specifications

Melting Point 54°C to 58°C
Boiling Point 171°C to 173°C (21 mmHg)
Flash Point 142°C (287°F)
Quantity 100 g
UN Number UN2923
Beilstein 472443
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-(1-Pyrrolidinyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'