missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)pyridine, 98%

Product Code. p-7027322
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11496876

Brand: Thermo Scientific Alfa Aesar B23989.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2456-81-7 | C9H12N2 | 148.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2456-81-7
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD00006415
InChI Key RGUKYNXWOWSRET-UHFFFAOYSA-N
Synonym 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
PubChem CID 75567
IUPAC Name 4-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=NC=C2

Specifications

Melting Point 54°C to 58°C
Boiling Point 171°C to 173°C (21 mmHg)
Flash Point 142°C (287°F)
Quantity 5 g
UN Number UN2923
Beilstein 472443
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-(1-Pyrrolidinyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.