missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10652253
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 143426-51-1
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD03659708
InChI Key LQQQPLUFBVYLRE-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol
PubChem CID 599478
IUPAC Name (4-pyrrol-1-ylphenyl)methanol
SMILES C1=CN(C=C1)C2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [4-(1H-Pyrrol-1-yl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'