missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Tetrazol-5-yl)phenol, 97%, Thermo Scientific™

Product Code. 11421490
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51517-88-5
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD05662709
InChI Key GDDQLGTXWIFSDI-UHFFFAOYSA-N
Synonym 4-1h-tetrazol-5-yl phenol, 5-4-hydroxyphenyl tetrazole, 5-4-hydroxyphenyl-1h-tetrazole, 4-1h-1,2,3,4-tetrazol-5-yl phenol, phenol, 4-1h-tetrazol-5-yl, 4-2h-tetrazol-5-yl phenol, 4-2h-tetrazol-5-yl-phenol, qoymnywfhzfxhr-uhfffaoysa-n, 4-2h-1,2,3,4-tetrazol-5-yl phenol, 4-1,2-dihydrotetrazol-5-ylidene cyclohexa-2,5-dien-1-one
PubChem CID 5413297
SMILES O=C1C=CC(C=C1)=C1NNN=N1

Specifications

Melting Point ∽240°C (decomposition)
Quantity 5 g
IUPAC Name 4-(2,5-dihydro-1H-1,2,3,4-tetrazol-5-ylidene)cyclohexa-2,5-dien-1-one
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 4-(1H-Tetrazol-5-yl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'