missing translation for 'onlineSavingsMsg'
Learn More

4-(2,3,5,6-Tetramethylphenylsulfonylamino)benzoic acid, 97%, Thermo Scientific™

Product Code. 15417905
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 730249-87-3
Molecular Formula C17H19NO4S
Molecular Weight (g/mol) 333.402
MDL Number MFCD03972197
InChI Key GXRBZEBVKOIXDU-UHFFFAOYSA-N
Synonym 4-2,3,5,6-tetramethylphenyl sulfonylamino benzoic acid, 4-2,3,5,6-tetramethylphenylsulfonylamino benzoic acid, 4-2,3,5,6-tetramethylbenzenesulfonamido benzoic acid, 4-2,3,5,6-tetramethylphenylsulfonamido benzoic acid, 4-2,3,5,6-tetramethylphenyl sulfonyl amino benzoic acid
PubChem CID 2411176
IUPAC Name 4-[(2,3,5,6-tetramethylphenyl)sulfonylamino]benzoic acid
SMILES CC1=CC(=C(C(=C1C)S(=O)(=O)NC2=CC=C(C=C2)C(=O)O)C)C

Specifications

Melting Point ∽230°C (decomposition)
Quantity 1 g
Formula Weight 333.41
Percent Purity 97%
Chemical Name or Material 4-(2,3,5,6-Tetramethylphenylsulfonylamino)benzoic acid
Correzione del contenuto del prodotto

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.