missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline, 97%
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
Descripción
It is an intermediate for the preparation of Elacridar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate for the preparation of Elacridar.
Solubility
Soluble in dichloromethane, ethyl acetate and methanol.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
It is an intermediate for the preparation of Elacridar.
Solubility
Soluble in dichloromethane, ethyl acetate and methanol.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Especificaciones
Especificaciones
| Quantity | 5 g |
| Solubility Information | Soluble in dichloromethane,ethyl acetate and methanol. |
| Formula Weight | 312.41 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido