missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals
£162.18 - £575.58
Chemical Identifiers
CAS | 30827-99-7 |
---|---|
Molecular Formula | C8H11ClFNO2S |
Molecular Weight (g/mol) | 239.689 |
InChI Key | WRDABNWSWOHGMS-UHFFFAOYSA-N |
Synonym | 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride |
PubChem CID | 186136 |
IUPAC Name | 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride |
SMILES | C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414822
|
MP Biomedicals
0219350380 |
100 mg |
£162.18
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424822
|
MP Biomedicals
0219350390 |
500 mg |
£575.58
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
30827-99-7 | |
239.689 | |
4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride | |
4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride |
C8H11ClFNO2S | |
WRDABNWSWOHGMS-UHFFFAOYSA-N | |
186136 | |
C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl |
Specifications
30827-99-7 | |
4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride | |
C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl | |
239.689 | |
239.7 |
C8H11ClFNO2S | |
WRDABNWSWOHGMS-UHFFFAOYSA-N | |
4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride | |
186136 | |
4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride |