missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 11401840
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13258-63-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00038045
InChI Key IDLHTECVNDEOIY-UHFFFAOYSA-N
Synonym 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine
PubChem CID 83275
IUPAC Name 2-pyridin-4-ylethanamine
SMILES NCCC1=CC=NC=C1

Specifications

Density 1.021
Boiling Point 70°C to 71°C (1 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.5395
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 4-(2-Aminoethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'