missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 15109223
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13258-63-4
Density 1.0210g/mL
Boiling Point 112.0°C to 113.0°C (12.0 mmHg)
Flash Point 100°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10N2
Linear Formula (C5H4N)CH2CH2NH2
Refractive Index 1.5403
MDL Number MFCD00038045
Quantity 1 g
Specific Gravity 1.021
Synonym 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine
InChI Key IDLHTECVNDEOIY-UHFFFAOYSA-N
SMILES NCCC1=CC=NC=C1
IUPAC Name 2-pyridin-4-ylethanamine
Molecular Weight (g/mol) 122.17
PubChem CID 83275
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 4-(2-Aminoethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(2-Aminoethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'