missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 203.08 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Bromoethyl)-3, 5-dimethyl-1H-pyrazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'