missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15480366
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957060-76-3
Molecular Formula C8H10BN3O3S
Molecular Weight (g/mol) 239.056
MDL Number MFCD09027244
InChI Key NVXNXTLWXKMYHB-UHFFFAOYSA-N
Synonym 2-4-boronobenzoyl hydrazinecarbothioamide, 4-2-carbamothioylhydrazinecarbonyl phenylboronic acid, 4-2-carbamothioylhydrazino carbonyl benzeneboronic acid, 4-carbamothioylaminocarbamoyl phenylboronic acid, 4-carbamothioylamino carbamoyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenylboronic
PubChem CID 44119176
IUPAC Name [4-[(carbamothioylamino)carbamoyl]phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NNC(=S)N)(O)O

Specifications

Melting Point 183°C to 184°C
Quantity 1 g
Formula Weight 239.06
Percent Purity 98%
Chemical Name or Material 4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'