missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride, 99%, Alfa Aesar™

Product Code. 15450166
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 913835-46-8
Molecular Formula C13H22BClN2O3
Molecular Weight (g/mol) 300.59
MDL Number MFCD09027220
InChI Key WQYCSVUEHAFRHV-UHFFFAOYSA-N
Synonym 4-2-diethylamino ethyl carbamoyl phenyl boronic acid hydrochloride, 4-2-n,n-diethylaminoethyl aminocarbonyl phenylboronic acid, hcl, 4-2-diethylamino ethyl carbamoyl benzeneboronic acid hydrochloride, 4-2-diethylamino ethyl carbamoyl phenylboronic acid hydrochloride, 4-2-diethylamino ethyl carbamoyl phenyl boronicacidhydrochloride, 4-2-n,n-diethylaminoethyl aminocarbonyl phenylboronic acid hcl, 4-2-diethylamino ethyl carbamoyl-phenyl boronic acid hydrochloride, 4-2-n,n-diethylaminoethyl aminocarbonyl benzeneboronic acid hydrochloride, 4-2-diethylamino ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1, boronic acid,b-4-2-diethylamino ethyl amino carbonyl phenyl-,hydrochloride 1:1
PubChem CID 44119574
IUPAC Name [4-[2-(diethylamino)ethylcarbamoyl]phenyl]boronic acid;hydrochloride
SMILES B(C1=CC=C(C=C1)C(=O)NCCN(CC)CC)(O)O.Cl

Specifications

Melting Point 65°C to 67°C
Quantity 250 mg
Formula Weight 300.59
Percent Purity 99%
Chemical Name or Material 4-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'