missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Furyl)but-3-en-2-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 50 g |
Formula Weight | 136.15 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Furyl)but-3-en-2-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'