missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)benzonitrile, Thermo Scientific™

Product Code. 10171132
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 69395-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
InChI Key RBSJBNYPTGMZIH-UHFFFAOYSA-N
Synonym 4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6
PubChem CID 2800815
IUPAC Name 4-(2-hydroxyethyl)benzonitrile
SMILES C1=CC(=CC=C1CCO)C#N

Specifications

Assay Percent Range 97%
Linear Formula CNC6H4(CH2)2OH
Quantity 250 mg
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'