missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyphenoxy)benzenesulfonyl chloride, 96%, Thermo Scientific™

Product Code. 15498325
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 690632-30-5
Molecular Formula C13H11ClO4S
Molecular Weight (g/mol) 298.74
MDL Number MFCD01631898
InChI Key DKNWPXFCLUKCQB-UHFFFAOYSA-N
PubChem CID 2794709
IUPAC Name 4-(2-methoxyphenoxy)benzenesulfonyl chloride
SMILES COC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Synonym 4-2-methoxyphenoxy benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-methoxyphenoxy, 4-2-methoxy-phenoxy-benzenesulfonyl chloride, 4-2-methoxyphenoxy benzene-1-sulfonyl chloride, 4-2-methoxyphenoxy benzenesulphonyl chloride, benzenesulfonylchloride,4-2-methoxyphenoxy, 4-2-methoxyphenoxy phenylsulfonyl chloride
Formula Weight 298.75
Percent Purity 96%
Chemical Name or Material 4-(2-Methoxyphenoxy)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'