missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methoxyphenyl)piperidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-2-methoxyphenyl piperidine, 4-2-methoxy-phenyl-piperidine, 4-2-methoxyphenyl-piperidine, piperidine, 4-2-methoxyphenyl, 2-methoxy-1-4-piperidyl benzene, pubchem17609, piperidine,4-2-methoxyphenyl |
Formula Weight | 191.27 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Methoxyphenyl)piperidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'