Learn More
[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol, 97%, Thermo Scientific™
£38.00 - £102.00
Chemical Identifiers
CAS | 857283-96-6 |
---|---|
Molecular Formula | C11H11NOS |
Molecular Weight (g/mol) | 205.275 |
MDL Number | MFCD07772836 |
InChI Key | RJHSPNFHEAFWDW-UHFFFAOYSA-N |
Synonym | 4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol |
PubChem CID | 7164608 |
IUPAC Name | [4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol |
SMILES | CC1=NC(=CS1)C2=CC=C(C=C2)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10177602
|
Thermo Scientific Maybridge
CC40309CB |
250 mg | Amber Glass Bottle |
£38.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10640895
|
Thermo Scientific Maybridge
CC40309DA |
1 g | Amber Glass Bottle |
£102.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
857283-96-6 | |
205.275 | |
RJHSPNFHEAFWDW-UHFFFAOYSA-N | |
7164608 | |
CC1=NC(=CS1)C2=CC=C(C=C2)CO |
C11H11NOS | |
MFCD07772836 | |
4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol | |
[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol |
Specifications
857283-96-6 | |
100.0 | |
C11H11NOS | |
4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol | |
CC1=NC(=CS1)C2=CC=C(C=C2)CO | |
205.275 | |
205.28 | |
[4-(2-Methyl-1, 3-thiazol-4-yl)phenyl]methanol |
96.5 | |
97% | |
MFCD07772836 | |
RJHSPNFHEAFWDW-UHFFFAOYSA-N | |
[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol | |
7164608 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning