missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-3-furylthio)-5-nonanone, 95%, Thermo Scientific™

Product Code. 11461860
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61295-50-9
Molecular Formula C14H22O2S
Molecular Weight (g/mol) 254.388
MDL Number MFCD08741453
InChI Key PEYZZTQOVLTVHN-UHFFFAOYSA-N
Synonym 4-2-methyl-3-furyl thio-5-nonanone, 4-2-methyl-3-furyl thio nonan-5-one, 4-2-methyl-3-furanyl thio-5-nonanone, 1,3-dipropylacetonyl 2-methyl-3-furyl sulfide, 4-2-methyl-furan-3-ylsulfanyl-nonan-5-one, 4-2-methyl-3-furylthio-5-nonanone, 4-2-methylfuran-3-yl sulfanylnonan-5-one, 5-nonanone, 4-2-methyl-3-furanyl thio, 4-2-methylfuran-3-yl sulfanyl nonan-5-one
PubChem CID 62182
IUPAC Name 4-(2-methylfuran-3-yl)sulfanylnonan-5-one
SMILES CCCCC(=O)C(CCC)SC1=C(OC=C1)C

Specifications

Refractive Index 1.4985
Quantity 100 mg
Formula Weight 254.39
Percent Purity 95%
Chemical Name or Material 4-(2-Methyl-3-furylthio)-5-nonanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'