missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide, Alfa Aesar™

Product Code. 11439790
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 902836-97-9
Molecular Formula C14H16N2O3S
Molecular Weight (g/mol) 292.35
MDL Number MFCD08061109
InChI Key AFXXBAMJRRMZDG-UHFFFAOYSA-N
Synonym 4-2-methylamino methyl phenoxy benzenesulfonamide, 4-2-methylaminomethyl phenoxy benzenesulfonamide, 4-2-methylaminomethyl-phenoxy-benzenesulfonamide, 4-2-methylamino methyl phenoxybenzenesulfonamid, 4-2-methylamino methyl phenoxy benzene-1-sulfonamide
PubChem CID 24208872
IUPAC Name 4-[2-(methylaminomethyl)phenoxy]benzenesulfonamide
SMILES CNCC1=CC=CC=C1OC1=CC=C(C=C1)S(N)(=O)=O

Specifications

Quantity 250 mg
Formula Weight 292.36
Chemical Name or Material 4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'