missing translation for 'onlineSavingsMsg'
Learn More
[4-(2-Morpholin-4-ylethoxy)phenyl]methanol, 97%, Thermo Scientific™ Product Code.: 10187602

[4-(2-Morpholin-4-ylethoxy)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10187602
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 10187602

Brand: Thermo Scientific Maybridge CC42109CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-76-8
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD07368553
InChI Key LZRLXLXSGAEQLA-UHFFFAOYSA-N
Synonym 4-2-morpholin-4-ylethoxy phenyl methanol, 4-2-morpholin-4-yl ethoxy phenyl methanol, 4-2-morpholinoethoxy phenyl methanol, benzenemethanol,4-2-4-morpholinyl ethoxy, 4-2-morpholin-4-yl-ethoxy-phenyl-methanol, 4-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7060580
IUPAC Name [4-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES OCC1=CC=C(OCCN2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material [4-(2-Morpholin-4-ylethoxy)phenyl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
[4-(2-Morpholin-4-ylethoxy)phenyl]methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.