Learn More
Memmert™ 4-20 mA Current Loop Interface for UF260
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF260V6
Product Code. 17529215
  • £384.00 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UF260 Oven
Current Loop Interface
4 to 20 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF260 > For Use With: UF260