Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Uf260Ts
Memmert™ Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF260TSV6
Product Code. 16354157
  • £384.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 4 to 20 mA current loop interface
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Current Loop Interface
4 to 20 mA