missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]benzaldehyde, 97%, Thermo Scientific™

Product Code. 10327174
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 26934-35-0
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD00011581
InChI Key ZYOHTXKOHWSJMZ-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy benzaldehyde, 4-3-dimethylaminopropoxy benzaldehyde, benzaldehyde, 4-3-dimethylamino propoxy, acmc-20ae0e, 4-3-dimethylamino propoxybenzaldehyde, 4-3-dimethylaminopropoxy-benzaldehyde, 4-3-dimethylamino-propoxy-benzaldehyde, 4-3-dimethylamino-propoxy benzaldehyde
PubChem CID 546932
IUPAC Name 4-[3-(dimethylamino)propoxy]benzaldehyde
SMILES CN(C)CCCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'