missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

£41.40 - £156.00

Chemical Identifiers

CAS 146440-15-5
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD09064993
InChI Key XYCJVOUBTBJYTL-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy benzonitrile, benzonitrile,4-3-dimethylamino propoxy, acmc-20n4ty, benzonitrile, 4-3-dimethylamino propoxy, 4-3-dimethylamino propoxy benzenecarbonitrile
PubChem CID 11858362
IUPAC Name 4-[3-(dimethylamino)propoxy]benzonitrile
SMILES CN(C)CCCOC1=CC=C(C=C1)C#N
View More Specs

Productos 2
Código de producto Marca Quantity Packaging Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Packaging Precio Cantidad y disponibilidad  
10356994
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC59516DA
1 g Amber Glass Bottle
£41.40
1g
Please to purchase this item. Need a web account? Register with us today!
10683235
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC59516DE
5 g Amber Glass Bottle
£156.00
5g
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Especificaciones
Show More
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
4-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado