missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Fluorophenylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15420466
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874288-05-8
Molecular Formula C13H11BFNO3
Molecular Weight (g/mol) 259.04
MDL Number MFCD09027204
InChI Key CVYDMRNYGKMSDI-UHFFFAOYSA-N
Synonym n-3-fluorophenyl 4-boronobenzamide, 4-3-fluorophenyl carbamoyl phenyl boronic acid, 4-borono-n-3-fluorophenyl benzamide, 4-3-fluorophenylcarbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl benzeneboronic acid, acmc-209qlr, n3-fluorophenyl 4-boronobenzamide
PubChem CID 44717609
IUPAC Name [4-[(3-fluorophenyl)carbamoyl]phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(=O)NC1=CC(F)=CC=C1

Specifications

Quantity 1 g
Formula Weight 259.04
Percent Purity 98%
Chemical Name or Material 4-(3-Fluorophenylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'