missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde, 90%, Thermo Scientific™

Product Code. 10082464
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10082464

Brand: Thermo Scientific Maybridge CC50904DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-27-7
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD09025865
InChI Key AEMXYVCSLPLJOW-UHFFFAOYSA-N
Synonym 4-3-methyl-1,2,4-oxadiazol-5-yl benzaldehyde, 3-methyl-5-4-formylphenyl-1,2,4-oxadiazole, benzaldehyde,4-3-methyl-1,2,4-oxadiazol-5-yl
PubChem CID 18525855
IUPAC Name 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
SMILES CC1=NOC(=N1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.19
Percent Purity 90%
Chemical Name or Material 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.