missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 204.18 |
Chemical Name or Material | 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction