missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-Thienyl)benzaldehyde, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Synonym | 4-thiophen-3-yl benzaldehyde, 4-3-thienyl benzaldehyde, 4-3-thienyl benzenecarbaldehyde, 4-thiophen-3-yl-benzaldehyde, benzaldehyde,4-3-thienyl, 4-thien-3-yl benzaldehyde, pubchem15939, thienylbenzenecarbaldehyde, 4-3-thiophenyl benzaldehyde, benzaldehyde, 4-3-thienyl |
Formula Weight | 188.25 |
Percent Purity | 97% |
Chemical Name or Material | 4-(3-Thienyl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'