missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 97%, Thermo Scientific™

Product Code. 10259282
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 364794-79-6
Molecular Formula C17H26BNO3
Molecular Weight (g/mol) 303.21
MDL Number MFCD04974052
InChI Key JOIXYIWXEYXHHG-UHFFFAOYSA-N
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 4-morpholinomethyl phenylboronic acid pinacol ester, 4-4-morpholinomethyl phenylboronic acid pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-methylmorpholino phenylboronic acid, pinacol ester, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-morpholinomethyl phenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-morpholin-4-ylmethyl phenyl-1,3,2-dioxaborolane
PubChem CID 2795502
IUPAC Name 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 303.21
Percent Purity 97%
Chemical Name or Material 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.