missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)-1H-indole, ≥97%, Thermo Scientific™

Product Code. 10397292
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-17-8
Molecular Formula C23H38BNO2Si
Molecular Weight (g/mol) 399.46
MDL Number MFCD05865142
InChI Key NGMKDRPSWHSLIM-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-indole, 1-tris isopropylsilyl-1h-indole-4-boronic acid, pinacol ester, 1-triisopropylsilyl-1h-indole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris isopropylsilyl-1h-indole, 1-triisopropylsilyl-1h-indole-4-boronic acid,pinacol ester, 1-triisopropylsilyl indole-4-boronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl indole, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-triisopropylsilanyl-1h-indole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tri propan-2-yl silyl-1h-indole, tri propan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indol-1-yl silane
PubChem CID 2795574
IUPAC Name tri(propan-2-yl)-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indol-1-yl]silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 399.46
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)-1H-indole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.