missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 97+%, Thermo Scientific Chemicals

Product Code. 10616884
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 269410-08-4
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.04
MDL Number MFCD03453063
InChI Key TVOJIBGZFYMWDT-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pyrazole-4-boronic acid pinacol ester, 4-pyrazoleboronic acid pinacol ester, 1h-pyrazole-4-boronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-4-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-1h-pyrazol-4-yl-1,3,2-dioxaborolane, pyrazole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 2-4-pyrazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2774010
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CC1(C)OB(OC1(C)C)C1=CNN=C1

Specifications

Melting Point 125.0°C to 130.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 194.04
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'