missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane, 97%

Product Code. 11986341 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11986341

Brand: Thermo Scientific Acros 431210010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1264198-72-2 | C12H24BNO2 | 225.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1264198-72-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 25.0°C to 27.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C12H24BNO2
MDL Number MFCD11506064
Quantity 1 g
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, 1-methylpiperidine-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl piperidine
InChI Key IMLCYAZBMGOKQS-UHFFFAOYSA-N
SMILES CN1CCC(CC1)B1OC(C)(C)C(C)(C)O1
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine
Molecular Weight (g/mol) 225.14
PubChem CID 45790007
Formula Weight 225.14
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.