missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Acetamidophenyl)-2-aminothiazole, 97%, Thermo Scientific™

Product Code. p-7030292
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11307459

Brand: Thermo Scientific Alfa Aesar H27854.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21674-96-4
Molecular Formula C11H11N3OS
Molecular Weight (g/mol) 233.289
MDL Number MFCD00207103
InChI Key VBBNSESFUHRMJU-UHFFFAOYSA-N
Synonym 4-4-acetamidophenyl-2-aminothiazole, n-4-2-amino-1,3-thiazol-4-yl phenyl acetamide, n-4-2-aminothiazol-4-yl phenyl acetamide, 2-amino-4-4-acetamidophenyl thiazole, n-4-4-amino-3,5-thiazolyl phenyl ethanamide, acetamide, n-4-2-amino-4-thiazolyl phenyl, acmc-1crp6, cbmicro_030947, aminothiazolylphenylacetamide
PubChem CID 519881
IUPAC Name N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C2=CSC(=N2)N

Specifications

Melting Point 248°C to 252°C
Odor Odorless
Quantity 5 g
Formula Weight 233.29
Percent Purity 97%
Chemical Name or Material 4-(4-Acetamidophenyl)-2-aminothiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.