missing translation for 'onlineSavingsMsg'
Learn More

4-(4-bromo-1,3-thiazol-2-yl)morpholine, 97%, Thermo Scientific™

Product Code. 10295574
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10295574

Brand: Thermo Scientific Maybridge CC62810DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1017781-60-0
Molecular Formula C7H9BrN2OS
Molecular Weight (g/mol) 249.126
MDL Number MFCD09607696
InChI Key XPFAGXZWVZAYCS-UHFFFAOYSA-N
Synonym 4-4-bromothiazol-2-yl morpholine, 4-4-bromo-1,3-thiazol-2-yl morpholine, morpholine, 4-4-bromo-2-thiazolyl, 4-4-bromo-thiazol-2-yl-morpholine, acmc-2097ww, 4-bromo-2-morpholinothiazole, morpholine,4-4-bromo-2-thiazolyl, 4-bromo-2-morpholin-4-yl-1,3-thiazole
PubChem CID 43811042
IUPAC Name 4-(4-bromo-1,3-thiazol-2-yl)morpholine
SMILES C1COCCN1C2=NC(=CS2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 249.13
Percent Purity 97%
Chemical Name or Material 4-(4-Bromo-1, 3-thiazol-2-yl)morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.