missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Bromophenyl)acetophenone, 97%, Thermo Scientific Chemicals

Artikelnummer. 11343257
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 5731-01-1
MDL Number MFCD00143242

Spezifikation

Melting Point 125°C to 127°C
Molecular Formula C14H11BrO
Quantity 25 g
Beilstein 2048590
Synonym 4-acetyl-4'-bromobiphenyl, 4'-4-bromophenyl acetophenone, 1-4-4-bromophenyl phenyl ethanone, 4'-p-bromophenyl acetophenone, 1-4'-bromo-1,1'-biphenyl-4-yl ethanone, 4-acetyl-4'-bromodiphenyl, 4-bromo-4'-acetylbiphenyl, ethanone, 1-4'-bromo 1,1'-biphenyl-4-yl, 1-4'-bromo 1,1'-biphenyl-4-yl ethanone, 1-4'-bromo 1,1'-biphenyl-4-yl ethan-1-one
InChI Key UUVKNCRMWPNBNM-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)Br
IUPAC Name 1-[4-(4-bromophenyl)phenyl]ethanone
Molecular Weight (g/mol) 275.15
PubChem CID 79806
Formula Weight 275.15
Percent Purity 97%
Chemical Name or Material 4'-(4-Bromophenyl)acetophenone
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'