missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Bromophenyl)butanoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 243.1 |
Percent Purity | 97% |
Chemical Name or Material | 4-(4-Bromophenyl)butanoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(4-Bromophenyl)butanoic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction