missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)butanoic acid, 97%, Thermo Scientific™

Product Code. 10651036
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35656-89-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00463170
InChI Key AGIIMNQWNPUJPT-UHFFFAOYSA-N
Synonym 4-4-bromophenyl butanoic acid, 4-4-bromophenyl butyric acid, benzenebutanoic acid, 4-bromo, 4-bromo-benzenebutanoic acid, pubchem16174, acmc-209iig, 4-bromobenzenebutanoic acid, benzenebutanoic acid,4-bromo, 4-4-bromo-phenyl-butyric acid
PubChem CID 252732
IUPAC Name 4-(4-bromophenyl)butanoic acid
SMILES C1=CC(=CC=C1CCCC(=O)O)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 243.1
Percent Purity 97%
Chemical Name or Material 4-(4-Bromophenyl)butanoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'